siRNA Details
| Gene Symbol: Gm5897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136397.1 | - | 1250 | |||
| BC147460.1 | - | 2 | 775 | 9056102 | ||
| BC147474.1 | - | 2 | 775 | 9056116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5897| Gene Symbol: Gm5897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136397.1 | - | 1250 | |||
| BC147460.1 | - | 2 | 775 | 9056102 | ||
| BC147474.1 | - | 2 | 775 | 9056116 | ||