siRNA Details
Gene Symbol: Ubtfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033793.3 | NP_001028965.1 | 3 | 2534 | ||
GenBank mRNA | AK139937.1 | - | 3 | 2536 | ||
BB289342.1 | - | 1 | 202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ubtfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033793.3 | NP_001028965.1 | 3 | 2534 | ||
GenBank mRNA | AK139937.1 | - | 3 | 2536 | ||
BB289342.1 | - | 1 | 202 |