siRNA Details
| Gene Symbol: Ccpg1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198789.1 | NP_001185718.1 | 1 | 94 | ||
| GenBank mRNA | AK145722.1 | - | 1 | 100 | ||
| BC048630.1 | - | 1 | 77 | |||
| BC099568.1 | - | 1 | 71 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccpg1os| Gene Symbol: Ccpg1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198789.1 | NP_001185718.1 | 1 | 94 | ||
| GenBank mRNA | AK145722.1 | - | 1 | 100 | ||
| BC048630.1 | - | 1 | 77 | |||
| BC099568.1 | - | 1 | 71 | |||