siRNA Details
| Gene Symbol: Ccpg1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198789.1 | NP_001185718.1 | 2 | 634 | ||
| GenBank mRNA | BC048630.1 | - | 2 | 617 | ||
| BC099568.1 | - | 2 | 611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccpg1os| Gene Symbol: Ccpg1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198789.1 | NP_001185718.1 | 2 | 634 | ||
| GenBank mRNA | BC048630.1 | - | 2 | 617 | ||
| BC099568.1 | - | 2 | 611 | |||