siRNA Details
| Gene Symbol: Fbxw18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033794.3 | NP_001028966.1 | 7, 8 | 1043 | ||
| GenBank mRNA | AK136230.1 | - | 7, 8 | 1047 | ||
| AK162167.1 | - | 7, 8 | 1028 | |||
| BC139081.1 | - | 7, 8 | 1043 | 8862103 | ||

Fbxw18
Genomic Map