siRNA Details
| Gene Symbol: Rhox3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145406.3 | NP_001138878.1 | 76 | |||
| GenBank mRNA | BC147475.1 | - | 76 | 9056117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox3g| Gene Symbol: Rhox3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145406.3 | NP_001138878.1 | 76 | |||
| GenBank mRNA | BC147475.1 | - | 76 | 9056117 | ||