siRNA Details
Gene Symbol: Prrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164275.2 | NP_001157747.1 | 4 | 456 | ||
NM_001289683.1 | NP_001276612.1 | 5 | 552 | |||
NM_027322.2 | NP_081598.1 | 4 | 483 | |||
GenBank mRNA | AK135895.1 | - | 5 | 547 | ||
CJ052035.1 | - | 4 | 481 | |||
CX242928.1 | - | 4 | 434 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
