siRNA Details
| Gene Symbol: Cyp26c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105201.1 | NP_001098671.1 | 6 | 1543 | ||
| GenBank mRNA | BC151098.1 | - | 6 | 1642 | 9088011 | |
| BC151106.1 | - | 6 | 1642 | 9088019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp26c1| Gene Symbol: Cyp26c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105201.1 | NP_001098671.1 | 6 | 1543 | ||
| GenBank mRNA | BC151098.1 | - | 6 | 1642 | 9088011 | |
| BC151106.1 | - | 6 | 1642 | 9088019 | ||