siRNA Details
| Gene Symbol: Gm13178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085536.1 | NP_001079005.1 | 4 | 749 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13178| Gene Symbol: Gm13178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085536.1 | NP_001079005.1 | 4 | 749 | ||