siRNA Details
| Gene Symbol: Ccdc42b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195094.1 | NP_001182023.1 | 4 | 322 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc42b| Gene Symbol: Ccdc42b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195094.1 | NP_001182023.1 | 4 | 322 | ||