siRNA Details
| Gene Symbol: Olfr455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081301.2 | NP_001074770.2 | 1 | 655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr455| Gene Symbol: Olfr455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081301.2 | NP_001074770.2 | 1 | 655 | ||