siRNA Details
| Gene Symbol: Vmn2r77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105188.1 | NP_001098658.1 | 4 | 1400 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r77| Gene Symbol: Vmn2r77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105188.1 | NP_001098658.1 | 4 | 1400 | ||
Set Membership: |
|