siRNA Details
| Gene Symbol: Dmrtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019872.1 | NP_063925.1 | 4 | 1350 | ||
| GenBank mRNA | AF020191.1 | - | 3 | 782 | ||
| AK132787.1 | - | 4 | 880 | |||
| AK161436.1 | - | 4 | 860 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmrtb1| Gene Symbol: Dmrtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019872.1 | NP_063925.1 | 4 | 1350 | ||
| GenBank mRNA | AF020191.1 | - | 3 | 782 | ||
| AK132787.1 | - | 4 | 880 | |||
| AK161436.1 | - | 4 | 860 | |||