siRNA Details
| Gene Symbol: Dmrtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019872.1 | NP_063925.1 | 3, 4 | 1007 | ||
| GenBank mRNA | AF020191.1 | - | 2, 3 | 439 | ||
| AK132787.1 | - | 3, 4 | 537 | |||
| AK161436.1 | - | 3, 4 | 517 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmrtb1| Gene Symbol: Dmrtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019872.1 | NP_063925.1 | 3, 4 | 1007 | ||
| GenBank mRNA | AF020191.1 | - | 2, 3 | 439 | ||
| AK132787.1 | - | 3, 4 | 537 | |||
| AK161436.1 | - | 3, 4 | 517 | |||