siRNA Details
| Gene Symbol: Kcnq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081142.1 | NP_001074611.1 | 6 | 870 | ||
| GenBank mRNA | AK040190.1 | - | 6 | 803 | ||
| AK163659.1 | - | 659 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq4| Gene Symbol: Kcnq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081142.1 | NP_001074611.1 | 6 | 870 | ||
| GenBank mRNA | AK040190.1 | - | 6 | 803 | ||
| AK163659.1 | - | 659 | ||||