siRNA Details
| Gene Symbol: Zcchc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033795.4 | NP_001028967.2 | 7 | 1295 | ||
| GenBank mRNA | AK136928.1 | - | 1 | 984 | ||
| AK141066.1 | - | 7 | 1297 | |||
| BC138958.1 | - | 1 | 571 | 8861980 | ||
| BC138959.1 | - | 1 | 571 | 8861981 | ||

Zcchc16
Genomic Map