siRNA Details
| Gene Symbol: Cyp2c11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019184.2 | NP_062057.2 | 2 | 280 | ||
| XM_008760364.1 | XP_008758586.1 | 1 | 108 | |||
| GenBank mRNA | BC088146.1 | - | 2 | 270 | 7369647 | |
| J02657.1 | - | 2 | 280 | |||
| U33173.1 | - | 2 | 280 | |||

Cyp2c11
Genomic Map