siRNA Details
| Gene Symbol: Zfp872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033813.4 | NP_001028985.1 | 4 | 2193 | ||
| GenBank mRNA | AK137156.1 | - | 4 | 2196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp872| Gene Symbol: Zfp872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033813.4 | NP_001028985.1 | 4 | 2193 | ||
| GenBank mRNA | AK137156.1 | - | 4 | 2196 | ||