siRNA Details
| Gene Symbol: Zfp551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033820.3 | NP_001028992.2 | 4 | 1297 | ||
| GenBank mRNA | AK137311.1 | - | 4 | 1298 | ||
| BC141160.1 | - | 4 | 1395 | 9055772 | ||
| BC145466.1 | - | 3 | 1271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp551| Gene Symbol: Zfp551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033820.3 | NP_001028992.2 | 4 | 1297 | ||
| GenBank mRNA | AK137311.1 | - | 4 | 1298 | ||
| BC141160.1 | - | 4 | 1395 | 9055772 | ||
| BC145466.1 | - | 3 | 1271 | |||