siRNA Details
| Gene Symbol: Gm6086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039219.2 | NP_001034308.1 | 8 | 3327 | ||
| GenBank mRNA | AK145447.1 | - | 8 | 3328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6086| Gene Symbol: Gm6086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039219.2 | NP_001034308.1 | 8 | 3327 | ||
| GenBank mRNA | AK145447.1 | - | 8 | 3328 | ||