siRNA Details
| Gene Symbol: Gm12695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081284.1 | NP_001074753.1 | 11 | 1575 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12695| Gene Symbol: Gm12695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081284.1 | NP_001074753.1 | 11 | 1575 | ||