siRNA Details
| Gene Symbol: Rhox3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040089.3 | NP_001035178.3 | 4 | 774 | ||
| GenBank mRNA | AY147207.1 | - | 667 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhox3f| Gene Symbol: Rhox3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040089.3 | NP_001035178.3 | 4 | 774 | ||
| GenBank mRNA | AY147207.1 | - | 667 | |||