siRNA Details
| Gene Symbol: Tmem170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163572.1 | NP_001157044.1 | 3 | 5808 | ||
| GenBank mRNA | AK150339.1 | - | 1 | 482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem170b| Gene Symbol: Tmem170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163572.1 | NP_001157044.1 | 3 | 5808 | ||
| GenBank mRNA | AK150339.1 | - | 1 | 482 | ||