siRNA Details
Gene Symbol: Tmem170b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163572.1 | NP_001157044.1 | 3 | 4195 | ||
GenBank mRNA | AK139239.1 | - | 1 | 1007 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem170b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163572.1 | NP_001157044.1 | 3 | 4195 | ||
GenBank mRNA | AK139239.1 | - | 1 | 1007 |