siRNA Details
| Gene Symbol: Vmn1r167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101562.1 | NP_001095032.1 | 1 | 808 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r167| Gene Symbol: Vmn1r167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101562.1 | NP_001095032.1 | 1 | 808 | ||