siRNA Details
| Gene Symbol: Kctd21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039039.3 | NP_001034128.2 | 2 | 2767 | ||
| GenBank mRNA | AK141547.1 | - | 2 | 2764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd21| Gene Symbol: Kctd21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039039.3 | NP_001034128.2 | 2 | 2767 | ||
| GenBank mRNA | AK141547.1 | - | 2 | 2764 | ||