siRNA Details
| Gene Symbol: 1700123I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165919.1 | NP_001159391.1 | 1, 2 | 180 | ||
| GenBank mRNA | AK007250.1 | - | 1, 2 | 186 | ||
| BC138005.1 | - | 1, 2 | 94 | 8861024 | ||
| BC145092.1 | - | 1, 2 | 94 | 9053640 | ||

1700123I01Rik
Genomic Map