siRNA Details
Gene Symbol: 1700123I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165919.1 | NP_001159391.1 | 11 | 1093 | ||
GenBank mRNA | AK007250.1 | - | 6 | 703 | ||
BC138005.1 | - | 11 | 1007 | 8861024 | ||
BC145092.1 | - | 11 | 1007 | 9053640 |
