siRNA Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294279.1 | NP_001281208.1 | 15 | 5767 | ||
| NM_178267.3 | NP_839998.2 | 15 | 5762 | |||
| GenBank mRNA | AK033589.1 | - | 1 | 1619 | ||
| AK035374.1 | - | 1 | 505 | |||
| BC041776.1 | - | 8 | 3229 | |||
| BC057076.1 | - | 1 | 1865 | |||
| BC088739.1 | - | 10 | 3392 | |||

Zfp827
Genomic Map