siRNA Details
| Gene Symbol: Krtap20-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163615.1 | NP_001157087.1 | 1 | 386 | ||
| GenBank mRNA | AK004483.1 | - | 1 | 387 | ||
| BY704708.1 | - | 1 | 387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap20-2| Gene Symbol: Krtap20-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163615.1 | NP_001157087.1 | 1 | 386 | ||
| GenBank mRNA | AK004483.1 | - | 1 | 387 | ||
| BY704708.1 | - | 1 | 387 | |||