siRNA Details
| Gene Symbol: Dok6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039173.1 | NP_001034262.1 | 5 | 518 | ||
| GenBank mRNA | AB221044.1 | - | 5 | 518 | ||
| AK140422.1 | - | 3 | 296 | |||
| BC138955.1 | - | 5 | 569 | 8861977 | ||
| BC138957.1 | - | 5 | 569 | 8861979 | ||

Dok6
Genomic Map