siRNA Details
| Gene Symbol: Nme9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165957.1 | NP_001159429.1 | 8 | 714 | ||
| Gene Symbol: 1600029I14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005560.1 | - | 1 | 455 | ||
| BC068205.1 | - | 1 | 473 | |||

Nme9
Genomic Map