siRNA Details
Gene Symbol: Nme9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165957.1 | NP_001159429.1 | 5 | 304 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nme9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165957.1 | NP_001159429.1 | 5 | 304 |