siRNA Details
| Gene Symbol: Fam43b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081672.2 | NP_001075141.1 | 1 | 2314 | ||
| GenBank mRNA | AK134570.1 | - | 1 | 2317 | ||
| BB677246.1 | - | 1 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam43b| Gene Symbol: Fam43b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081672.2 | NP_001075141.1 | 1 | 2314 | ||
| GenBank mRNA | AK134570.1 | - | 1 | 2317 | ||
| BB677246.1 | - | 1 | 377 | |||