siRNA Details
| Gene Symbol: Zfp951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039231.3 | NP_001034320.3 | 5 | 2672 | ||
| GenBank mRNA | AK132470.1 | - | 6 | 1500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp951| Gene Symbol: Zfp951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039231.3 | NP_001034320.3 | 5 | 2672 | ||
| GenBank mRNA | AK132470.1 | - | 6 | 1500 | ||