siRNA Details
| Gene Symbol: Gm14322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243903.1 | NP_001230832.1 | 5 | 2297 | ||
| GenBank mRNA | BC092145.1 | - | 6 | 1391 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14322| Gene Symbol: Gm14322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243903.1 | NP_001230832.1 | 5 | 2297 | ||
| GenBank mRNA | BC092145.1 | - | 6 | 1391 | ||