siRNA Details
| Gene Symbol: Vmn2r90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104539.1 | NP_001098009.1 | 1, 2 | 204 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r90| Gene Symbol: Vmn2r90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104539.1 | NP_001098009.1 | 1, 2 | 204 | ||
Set Membership: |
|