siRNA Details
| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081678.1 | NP_001075147.1 | 6 | 2426 | ||
| GenBank mRNA | AK036691.1 | - | 7 | 2693 | ||
| BC066073.1 | - | 6 | 2839 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp800| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081678.1 | NP_001075147.1 | 6 | 2426 | ||
| GenBank mRNA | AK036691.1 | - | 7 | 2693 | ||
| BC066073.1 | - | 6 | 2839 | |||