siRNA Details
| Gene Symbol: Fam196a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143802.1 | NP_001137274.1 | 4 | 3592 | ||
| GenBank mRNA | AK046846.1 | - | 3 | 2308 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam196a| Gene Symbol: Fam196a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143802.1 | NP_001137274.1 | 4 | 3592 | ||
| GenBank mRNA | AK046846.1 | - | 3 | 2308 | ||