siRNA Details
| Gene Symbol: Gm14092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037929.2 | NP_001033018.1 | 3 | 951 | ||
| GenBank mRNA | AK133124.1 | - | 3 | 952 | ||
| BC147590.1 | - | 3 | 927 | 9056580 | ||
| BC147611.1 | - | 3 | 927 | 9056601 | ||

Gm14092
Genomic Map