siRNA Details
| Gene Symbol: Gm6760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177377.1 | NP_001170848.1 | 2 | 372 | ||
| GenBank mRNA | BC048649.1 | - | 2 | 267 | ||
| BU939216.1 | - | 2 | 373 | |||
| CN841342.1 | - | 2 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6760| Gene Symbol: Gm6760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177377.1 | NP_001170848.1 | 2 | 372 | ||
| GenBank mRNA | BC048649.1 | - | 2 | 267 | ||
| BU939216.1 | - | 2 | 373 | |||
| CN841342.1 | - | 2 | 283 | |||