siRNA Details
| Gene Symbol: Vmn2r100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104562.1 | NP_001098032.1 | 4 | 1354 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r100| Gene Symbol: Vmn2r100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104562.1 | NP_001098032.1 | 4 | 1354 | ||
Set Membership: |
|