siRNA Details
| Gene Symbol: Gm6878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037931.3 | NP_001033020.1 | 4 | 1519 | ||
| GenBank mRNA | AK134891.1 | - | 4 | 1521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6878| Gene Symbol: Gm6878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037931.3 | NP_001033020.1 | 4 | 1519 | ||
| GenBank mRNA | AK134891.1 | - | 4 | 1521 | ||