siRNA Details
| Gene Symbol: Gm6890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099306.1 | NP_001092776.1 | 3 | 678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6890| Gene Symbol: Gm6890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099306.1 | NP_001092776.1 | 3 | 678 | ||