siRNA Details
| Gene Symbol: Hs3st4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252072.1 | NP_001239001.1 | 2 | 1510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st4| Gene Symbol: Hs3st4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252072.1 | NP_001239001.1 | 2 | 1510 | ||