siRNA Details
| Gene Symbol: Zfp953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038651.3 | NP_001033740.1 | 4 | 3278 | ||
| GenBank mRNA | AK143165.1 | - | 4 | 3279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp953| Gene Symbol: Zfp953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038651.3 | NP_001033740.1 | 4 | 3278 | ||
| GenBank mRNA | AK143165.1 | - | 4 | 3279 | ||