siRNA Details
Gene Symbol: Fam124a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243857.1 | NP_001230786.1 | 4 | 3514 | ||
GenBank mRNA | AK137188.1 | - | 1 | 1266 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam124a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243857.1 | NP_001230786.1 | 4 | 3514 | ||
GenBank mRNA | AK137188.1 | - | 1 | 1266 |