siRNA Details
| Gene Symbol: Fam124a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243857.1 | NP_001230786.1 | 4 | 3286 | ||
| GenBank mRNA | AK137188.1 | - | 1 | 1038 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam124a| Gene Symbol: Fam124a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243857.1 | NP_001230786.1 | 4 | 3286 | ||
| GenBank mRNA | AK137188.1 | - | 1 | 1038 | ||