siRNA Details
| Gene Symbol: Ctxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134697.1 | NP_001128169.1 | 2 | 792 | ||
| GenBank mRNA | AK042789.1 | - | 2 | 752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctxn3| Gene Symbol: Ctxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134697.1 | NP_001128169.1 | 2 | 792 | ||
| GenBank mRNA | AK042789.1 | - | 2 | 752 | ||