siRNA Details
| Gene Symbol: Sult2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081325.2 | NP_001074794.1 | 2 | 276 | ||
| GenBank mRNA | BC147210.1 | - | 2 | 276 | 9056256 | |
| BC172008.1 | - | 2 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult2a6| Gene Symbol: Sult2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081325.2 | NP_001074794.1 | 2 | 276 | ||
| GenBank mRNA | BC147210.1 | - | 2 | 276 | 9056256 | |
| BC172008.1 | - | 2 | 276 | |||